Catalyst Precision Stopped Assemblies for FH100 Series Pumps
Start pumping with little effort - preconfigured Links for quick and easy setup
  • Available in Platinum-Cured Silicone, TPEs, and PVC
  • Save time with ready-to-use links / assemblies
  • Ensure the optimal amount of tubing is in the pump head
  • FH100 and FH100D utilize tubing with a wall thickness of 1.6 mm
  • FH100X and FH100DX utilize tubing with a wall thickness of 2.4 mm
To complete your Catalyst® pumping system, order two-stop color-coded and extension tubing. Tubing comes in various formulations for almost any pumping application.
The stops on the two-stop tubing are color-coded according to tubing ID.
Order extension tubing separately. For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.
$232.00 - $2,099.00USD / Pkg of 1
40 variations of this product are available.
See All Product Options Below
40 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing Formulation Tubing ID (mm) Wall Thickness (mm)
Availability Pricing
COMPARE
PharMed® BPT 0.8 1.6
$296.00USD / Pkg of 1
COMPARE
PharMed® BPT 1.6 1.6
$301.00USD / Pkg of 1
COMPARE
PharMed® BPT 3.2 1.6
$308.00USD / Pkg of 1
COMPARE
PharMed® BPT 6.4 1.6
$316.00USD / Pkg of 1
COMPARE
PharMed® BPT 8 1.6
$361.00USD / Pkg of 1
COMPARE
Tygon® A-60-F 3.2 1.6
$274.00USD / Pkg of 1
COMPARE
Tygon® A-60-F 4.8 1.6
$274.00USD / Pkg of 1
COMPARE
Tygon® A-60-F 6.4 1.6
$2,099.00USD / Pkg of 1
COMPARE
Tygon® A-60-F 8 1.6
$303.00USD / Pkg of 1
COMPARE
Tygon® A-60-F 6.4 2.4
$316.00USD / Pkg of 1
COMPARE
Tygon® A-60-F 8 2.4
$321.00USD / Pkg of 1
COMPARE
Tygon® A-60-F 9.5 2.4
$342.00USD / Pkg of 1
COMPARE
Tygon® E-Food (B-44-4X) 1.6 1.6
$246.00USD / Pkg of 12
COMPARE
Tygon® E-Food (B-44-4X) 4.8 1.6
$265.00USD / Pkg of 12
COMPARE
Tygon® E-Food (B-44-4X) 6.4 1.6
$268.00USD / Pkg of 12
COMPARE
Tygon® E-Food (B-44-4X) 9.5 2.4
$397.00USD / Pkg of 12
COMPARE
Platinum-Cured Silicone 0.8 1.6
$344.00USD / Pkg of 12
COMPARE
Platinum-Cured Silicone 6.4 1.6
$401.00USD / Pkg of 12
COMPARE
Platinum-Cured Silicone 4.8 2.4
$369.00USD / Pkg of 12
COMPARE
Platinum-Cured Silicone 6.4 2.4
$418.00USD / Pkg of 12
Showing 1 - 20 of 40