Catalyst Pump Coiled Tubing For FH100, FH100D, FH100X and FH100DX Pumps
Four formulations available to cover most chemical compatible applications
  • To help simplify choices, a platinum-cured silicone, two TPEs and a long life PVC tubing material are offered.
  • Standard 15 meter lengths
  • Available in both thin and thick wall configurations.
  • FH100 and FH100D utilize tubing with a wall thickness of 1.6 mm.
  • FH100X and FH100DX utilize tubing with a wall thickness of 2.4 mm.
Choose from a wide range of tubing options for your Catalyst pumps.
$46.50 - $504.00USD / Pkg of 1
28 variations of this product are available.
See All Product Options Below
28 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing Formulation Tubing ID (mm) Wall Thickness (mm)
Availability Pricing
COMPARE
PharMed® BPT 1.6 1.6
$266.00USD / Pkg of 1
COMPARE
PharMed® BPT 3.2 1.6
$263.00USD / Pkg of 1
COMPARE
PharMed® BPT 4.8 1.6
$269.00USD / Pkg of 1
COMPARE
PharMed® BPT 6.4 1.6
$318.00USD / Pkg of 1
COMPARE
PharMed® BPT 8 1.6
$331.00USD / Pkg of 1
COMPARE
Tygon® A-60-F 1.6 1.6
$71.00USD / Pkg of 1
COMPARE
Tygon® A-60-F 4.8 1.6
$108.00USD / Pkg of 1
COMPARE
Tygon® A-60-F 6.4 1.6
$128.00USD / Pkg of 1
COMPARE
Tygon® A-60-F 8 1.6
$151.00USD / Pkg of 1
COMPARE
Tygon® E-Food (B-44-4X) 3.2 1.6
$66.00USD / Pkg of 1
COMPARE
Tygon® E-Food (B-44-4X) 4.8 1.6
$87.00USD / Pkg of 1
COMPARE
Tygon® E-Food (B-44-4X) 8 1.6
$132.00USD / Pkg of 1
COMPARE
Tygon® E-Food (B-44-4X) 6.4 2.4
$182.00USD / Pkg of 1
COMPARE
Tygon® E-Food (B-44-4X) 8 2.4
$201.00USD / Pkg of 1
COMPARE
Tygon® E-Food (B-44-4X) 9.5 2.4
$268.00USD / Pkg of 1
COMPARE
Platinum-Cured Silicone 1.6 1.6
$145.00USD / Pkg of 1
COMPARE
Platinum-Cured Silicone 4.8 1.6
$236.00USD / Pkg of 1
COMPARE
Platinum-Cured Silicone 6.4 1.6
$232.00USD / Pkg of 1
COMPARE
Platinum-Cured Silicone 6.4 2.4
$379.00USD / Pkg of 1
COMPARE
Platinum-Cured Silicone 9.5 2.4
$504.00USD / Pkg of 1
Showing 1 - 20 of 28