Catalyst Pump Tubing Links / Assemblies for FH15, FH30 and FH100M Pumps
Start pumping with little effort - preconfigured Links for quick and easy setup
  • Four formulations offer wide range of chemical compatibility
  • Save time with already assembled links
  • Use with the Catalyst extenstion coils / tubing to complete your system
  • FH15 and FH30 utilize the stopped assemblies that have an ID of: 0.8, 1.6, 3.2, and 4.8 mm
  • FH100M (with the large cartridges) can use assemblies with an ID of: 0.8, 1.6, 3.2, 4.8, and 6.4 mm
  • FH100M (with the small cartridges) can use assemblies with an ID of: 0.16, 0.25, 0.89, 1.42, 2.06, and 2.79 mm
To complete your Catalyst® pumping system, order two-stop color-coded and extension tubing. Tubing comes in various formulations for almost any pumping application.
The stops on the two-stop tubing are color-coded according to tubing ID.
Order extension tubing separately. For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.
$81.00 - $176.00USD / Pkg of 1
31 variations of this product are available.
See All Product Options Below
31 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing Formulation Tubing ID (mm) Wall Thickness (mm)
Availability Pricing
COMPARE
Platinum-Cured Silicone 4.8 2.4
$176.00USD / Pkg of 1
COMPARE
Platinum-Cured Silicone 6.4
$176.00USD / Pkg of 1
COMPARE
PharMed® BPT 0.8 1.6
$98.50USD / Pkg of 1
COMPARE
PharMed® BPT 1.6 1.6
$116.00USD / Pkg of 1
COMPARE
PharMed® BPT 4.8 2.4
$99.50USD / Pkg of 1
COMPARE
Tygon® A-60-F 0.8 1.6
$81.00USD / Pkg
COMPARE
Tygon® A-60-F 1.6 1.6
$81.00USD / Pkg
COMPARE
Tygon® A-60-F 4.8 2.4
$81.50USD / Pkg
COMPARE
Tygon® A-60-F 6.4 2.4
$81.50USD / Pkg
COMPARE
Tygon® E-Food (B-44-4X) 1.6 1.6
$90.50USD / Pkg of 1
COMPARE
Tygon® E-Food (B-44-4X) 3.2 3.2
$90.50USD / Pkg of 1
COMPARE
Platinum-Cured Silicone 1.42 0.86
$91.00USD / Pkg of 6
COMPARE
Platinum-Cured Silicone 2.79 0.86
$91.00USD / Pkg of 6
COMPARE
PharMed® BPT 0.25 0.91
$159.00USD / Pkg of 12
COMPARE
PharMed® BPT 1.42 0.86
$159.00USD / Pkg of 12
COMPARE
PharMed® BPT 2.79 0.86
$159.00USD / Pkg of 12
COMPARE
Tygon® E-Food (B-44-4X) 0.19 0.91
$85.00USD / Pkg of 12
COMPARE
Tygon® E-Food (B-44-4X) 0.89 0.86
$85.00USD / Pkg of 12
COMPARE
Tygon® E-Food (B-44-4X) 2.06 0.86
$82.50USD / Pkg of 12
COMPARE
Tygon® E-Food (B-44-4X) 2.79 0.86
$85.50USD / Pkg of 12
Showing 1 - 20 of 31