This product is now discontinued, View Related Products

Caution: Watch Your Step Signs

These signs are intended to indicate potential, type, and degree of the hazard that may lead to accidental injury or property damage. Signs define specific hazards by conveying a clear, easily understood message. The sign headers and colors used in accident prevention safety signs are specified by OSHA and ANSI to designate the severity of the hazard. Each color has a specific role.

CAUTION (yellow): Indicates a potentially hazardous situation which, if not avoided, may result in minor or moderate injury.
Call For Price

6 variations of this product are available.

See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Height (in) Material Width (in)
Availability Pricing
Accuform MSTF645VS Safety Sign, Caution - Watch Your Step, 7" X 10", Adhesive Vinyl EW-50710-38 Mfr # MSTF645VS
COMPARE
7 Adhesive Vinyl 10
Call For Price
Accuform MSTF645VP Safety Sign, Caution - Watch Your Step, 7" X 10", Plastic EW-50710-39 Mfr # MSTF645VP
COMPARE
7 Plastic 10
Call For Price
Accuform MSTF645VA Safety Sign, Caution - Watch Your Step, 7" X 10", Aluminum EW-50710-40 Mfr # MSTF645VA
COMPARE
7 Aluminum 10
Call For Price
Accuform MCSPC37VS Safety Sign, Caution - Watch Your Step, 10" X 14", Adhesive Vinyl EW-51014-38 Mfr # MCSPC37VS
COMPARE
10 Adhesive Vinyl 14
Call For Price
Accuform MCSPC37VP Safety Sign, Caution - Watch Your Step, 10" X 14", Plastic EW-51014-39 Mfr # MCSPC37VP
COMPARE
10 Plastic 14
Call For Price
Accuform MCSPC37VA Safety Sign, Caution - Watch Your Step, 10" X 14", Aluminum EW-51014-40 Mfr # MCSPC37VA
COMPARE
10 Aluminum 14
Call For Price
Showing 1- 6 of 6

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat