Centrifuge Tube Ice Tray, polypropylene
For temperature stabilized benchtop ice storage
This heavy duty, all polypropylene rack provides efficient and extended cooling of twenty-eight 15 ml or 50 ml centrifuge tubes in a chilled bath. The deep bottom tray holds approximately 2 liters of ice flakes and water so that tubes can rest on the bottom of the tray for maximum stability and exposure to the bath. A flat perimeter rim provides space for labeling tape. The rack is steam autoclavable at 121°C (250°F) and stackable with identical racks when full or empty. To further protect tubes during transport or storage while also maintaining stackability, an optional polypropylene cover is available (order separately).
$36.00 - $95.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Material Description
Availability Pricing
Scienceware F18905-0050 Centrifuge Tube Ice Rack (holds 28 X 50 mL Tubes) EW-06274-04 Mfr # F18905-0050
COMPARE
Polypropylene Centrifuge Tube Ice Rack (holds 28 X 50 mL Tubes)
$95.00USD / Each
Scienceware F18905-0051 Centrifuge Tube Ice Rack Cover, polypropylene EW-06274-06 Mfr # F18905-0051
COMPARE
Polypropylene Centrifuge Tube Ice Rack Cover, polypropylene
$36.00USD / Each