This product is now discontinued, View Related Products
CheckPoint O2 and CO2/O2 Analyzers
Measure gas levels in modified atmosphere packages (MAP) in just 10 seconds
  • Internal memory function stores up to 10 tests
  • Battery powered for up to 2400 measurements
Compact, hand-held CheckPoint analyzer packs excellent functionality in a portable, easy-to-use instrument. Simply use probe assembly to penetrate package to be sampled. Powerful, internal pump draws 15mL sample and displays gas concentration level in seconds on bright, 3-digit LED display. Memory function allows you to store up to 10 readings to be recorded later. Auto-calibration feature ensures quick calibration at the touch of a button. Analyzer will power off automatically after one minute of inactivity to save battery life. Display hold option enables you to lock the display for easy readability.
Analyzers are encased in a rugged, anodized aluminum housing with IP53 rating for impact and splash resistance. Integrated needle holder prevents accidental sticks when probe is not in use. Select from single O2 or combined O2/CO2 model to suit your application.
What's included:sampling probe assembly, 10 septa, 10 repl. needles and filters, four AA batteries, and a manual.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Gases Measured
Availability Pricing
Mocon 200547 Headspace Analyzer, Handheld, 02 EW-86490-00 Mfr # 200547
COMPARE
O2
Call For Price
Mocon 200548 Headspace Analyzer, Handheld, Co2/02 EW-86490-02 Mfr # 200548
COMPARE
CO2, O2