Chem-Tec Adjustable Mini Low-Flow Switches
Adjustable set points trigger SPST contacts rated for 70 VA
  • Choose from brass or TFE materials
Mini Low-Flow Switches are configured to open the SPST contact when flow goes beyond the set point or stops. Typical applications include gas/liquid sampling; chemical injection; pollution control monitoring; atmospheric furnaces; and process systems. Choose from a brass switch (32929-00) for pressures up to 1500 psi or a switch made from PTFE (32929-02) for good chemical compatibility. Wetted materials are brass, epoxy, and Viton® for model 32929-00; PTFE for model 32929-02. Repeatability is ±2%. Measure 1-¼''W x 2-5/8''H x ¾''D.
$279.00 - $337.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Media Type Wetted Materials Description
Availability Pricing
Chem-Tec 125-BNO Adjustable Mini Low-Flow Switch for Liquids & Gases, Brass EW-32929-00 Mfr # 125-BNO
COMPARE
Adjustable Mini Low-Flow Switch for Liquids & Gases, Brass
$279.00USD / Each
Chem-Tec 125-TNO Adjustable Mini Low-Flow Switch for Liquids & Gases, PTFE EW-32929-02 Mfr # 125-TNO
COMPARE
Adjustable Mini Low-Flow Switch for Liquids & Gases, PTFE
$337.00USD / Each