Chem-Tec Industrial Adjustable Flow Switches
Adjustable set points trigger SPST contacts rated for 70 VA
  • Choose from brass or TFE materials
Industrial Flow Switches are normally closed (NC) but can be wired for normally open (NO). Typical applications include lubrication, process, fire control, and cooling systems; heat pumps; hydraulic lifts; and water treatment.
Choose a brass model with a ¼'', 3/8'', ½'', or ¾'' NPT(F) connection. Repeatability is ±2%. Note: Back pressure of 5 psi is required for gas applications. Wetted materials are brass, 316 SS, epoxy, and Viton for all models. Models 32929-20, -22, and -24 measure 3-¼''W x 3-1/4''H x 1''D; model 32929-26 measures 4-¼''W x 4''H x 1''D. Shpg wt 4 lb (1.9 kg).
$331.00 - $589.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Media Type Pipe Size Description
Availability Pricing
Chem-Tec Adjustable Industrial Flow Switch for Liquids & Gases, 1/4" NPT(F) Conn EW-32929-20 Mfr # FAV250BWNC
COMPARE
Adjustable Industrial Flow Switch for Liquids & Gases, 1/4" NPT(F) Conn
$331.00USD / Each
Chem-Tec Adjustable Industrial Flow Switch for Liquids & Gases, 3/8" NPT(F) Conn EW-32929-22 Mfr # FAV375BWNC
COMPARE
Adjustable Industrial Flow Switch for Liquids & Gases, 3/8" NPT(F) Conn
$353.00USD / Each
Chem-Tec Adjustable Industrial Flow Switch for Liquids & Gases, 1/2" NPT(F) Conn EW-32929-24 Mfr # FAV500BWNC
COMPARE
Adjustable Industrial Flow Switch for Liquids & Gases, 1/2" NPT(F) Conn
$382.00USD / Each
Chem-Tec Adjustable Industrial Flow Switch for Liquids & Gases, 3/4" NPT(F) Conn EW-32929-26 Mfr # FAV750BWNC
COMPARE
Adjustable Industrial Flow Switch for Liquids & Gases, 3/4" NPT(F) Conn
$589.00USD / Each