Chemfluor 367 Tubing 17202
Applications: Pharmaceutical, semiconductor, electrical, and environmental applications. Excellent for processes dependent on maintaining fluid integrity and high purity.
Characteristics: Exceptionally smooth surface, very low extractables with deionized water and acids, excellent chemical resistance and mechanical properties, low counts of trace metals, superior clarity.
Certification: FDA compliant.
Temperature range: -400 to 450°F (-240 to 232°C)
Sterilize: With ethylene oxide, autoclave, or dry heat.
$96.50 - $632.00USD / Pkg of 1
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
ChemFluor TSC367-0125-031-50 367 Tubing, 379 psi at 73°F
Buy More Save More
EW-95638-00 Mfr # TSC367-0125-031-50
COMPARE
379 1/16 1/8
$96.50USD / Pkg of 1
ChemFluor TSC367-0250-062-50 367 Tubing, 383 psi at 73°F
Buy More Save More
EW-95638-01 Mfr # TSC367-0250-062-50
COMPARE
383 1/8 1/4
$324.00USD / Pkg of 1
ChemFluor TSC367-0250-047-50 367 Tubing, 266 psi at 73°F
Buy More Save More
EW-95638-02 Mfr # TSC367-0250-047-50
COMPARE
266 5/32 1/4
$281.00USD / Pkg of 1
ChemFluor TSC367-0250-031-50 367 Tubing, 163 psi at 73°F
Buy More Save More
EW-95638-03 Mfr # TSC367-0250-031-50
COMPARE
163 3/16 1/4
$208.00USD / Pkg of 1
ChemFluor TSC367-0312-031-50 367 Tubing, 129 psi at 73°F
Buy More Save More
EW-95638-04 Mfr # TSC367-0312-031-50
COMPARE
129 1/4 5/16
$259.00USD / Pkg of 1
ChemFluor TSC367-0375-062-50 367 Tubing, 230 psi at 73°F
Buy More Save More
EW-95638-05 Mfr # TSC367-0375-062-50
COMPARE
230 1/4 3/8
$495.00USD / Pkg of 1
ChemFluor TSC367-0500-062-50 367 Tubing, 164 psi at 73°F
Buy More Save More
EW-95638-06 Mfr # TSC367-0500-062-50
COMPARE
164 3/8 1/2
$632.00USD / Pkg of 1
Showing 1- 7 of 7