Chemfluor ETFE Tubing
Tubing is translucent and slightly flexible. Tubing is versatile and can be used in several applications, including lab, chemical, electrical, mechanical, and automotive applications. Ideal for biochromatography-will not denature sensitive biomolecules. ETFE exhibits excellent mechanical resistance and is more chemically inert than SS tubing. Withstands pressures up to 3000 psi and can be used continuously at temperatures up to 300°>F (149°>C). The resin meets ASTM standard D3159 and may be sterilized by autoclaving or ethylene oxide. Pack of 50 feet, unless noted otherwise.
$173.00 - $436.00USD / Pkg of 1
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
ChemFluor TSTZ83-0062-021-50 ETFE tubing, 1/50 x 1/16", 50 ft/pk
Buy More Save More
EW-06488-07 Mfr # TSTZ83-0062-021-50
COMPARE
975 1/50 1/16
$206.00USD / Pkg of 1
ChemFluor TSTZ83-0062-023-50 ETFE tubing, 0.016 x 1/16", 50 ft/pk
Buy More Save More
EW-06488-08 Mfr # TSTZ83-0062-023-50
COMPARE
975 1/64 1/16
$229.00USD / Pkg of 1
ChemFluor TSTZ83-0125-016-50 ETFE tubing, 3/32 x 1/8", 50 ft/pk
Buy More Save More
EW-06488-09 Mfr # TSTZ83-0125-016-50
COMPARE
975 3/32 1/8
$173.00USD / Pkg of 1
ChemFluor TSTZ83-0125-031-50 ETFE tubing, 1/16 x 1/8", 50 ft/pk
Buy More Save More
EW-06488-10 Mfr # TSTZ83-0125-031-50
COMPARE
975 1/16 1/8
$262.00USD / Pkg of 1
ChemFluor TSTZ83-0250-031-50 ETFE tubing, 3/16 x 1/4", 50 ft/pk
Buy More Save More
EW-06488-11 Mfr # TSTZ83-0250-031-50
COMPARE
455 3/16 1/4
$229.00USD / Pkg of 1
ChemFluor TSTZ83-0250-062-50 ETFE tubing, 1/8 x 1/4", 50 ft/pk
Buy More Save More
EW-06488-12 Mfr # TSTZ83-0250-062-50
COMPARE
455 1/8 1/4
$353.00USD / Pkg of 1
ChemFluor TSTZ83-0312-040-50 ETFE tubing, 0.232 x 5/16", 50 ft/pk
Buy More Save More
EW-06488-13 Mfr # TSTZ83-0312-040-50
COMPARE
290 15/64 5/16
$298.00USD / Pkg of 1
ChemFluor TSTZ83-0375-048-50 ETFE tubing, 0.278 x 3/8", 50 ft/pk
Buy More Save More
EW-06488-14 Mfr # TSTZ83-0375-048-50
COMPARE
290 9/32 3/8
$309.00USD / Pkg of 1
ChemFluor TSTZ83-0500-062-50 ETFE tubing, 3/8 x 1/2", 50 ft/pk
Buy More Save More
EW-06488-15 Mfr # TSTZ83-0500-062-50
COMPARE
215 3/8 1/2
$436.00USD / Pkg of 1
Showing 1- 9 of 9