Chemfluor FEP Tubing 18616
Applications: Transfer of abrasive fluids; pharmaceutical processes.
Characteristics: Good chemical resistance at a low cost, temperature resistant, low coefficient of friction, semi-flexible. Transparent.
Certification:FDA compliant. Meets USP 23 Class VI.
Temperature range:–454 to 400°F (–270 to 205°C
Sterilize:With autoclave, ethylene oxide, or use dry heat.
$69.50 - $1,622.00USD / Pkg of 1
21 variations of this product are available.
See All Product Options Below
21 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
ChemFluor TSFE14-0125-031-50 FEP Tubing, 213 psi @ 70F, 1/16" x 1/8", 50ft/pk
Buy More Save More
EW-96001-02 Mfr # TSFE14-0125-031-50
COMPARE
213 1/16 1/8
$69.50USD / Pkg of 1
ChemFluor TSFE14-0187-062 FEP Tubing, 283 psi @ 70F, 1/16 X 3/16, 50'
Buy More Save More
EW-96001-04 Mfr # TSFE14-0187-062
COMPARE
283 1/16 3/16
$418.00USD / Pkg of 1
ChemFluor TSFE14-0187-031 FEP tubing, 1/8 x 3/16, pack of 50'
Buy More Save More
EW-96001-08 Mfr # TSFE14-0187-031
COMPARE
142 1/8 3/16
$127.00USD / Pkg of 1
ChemFluor TSFE14-0250-062 FEP Tubing, 213 psi @ 70F, 1/8" x 1/4", 50ft/pk
Buy More Save More
EW-96001-10 Mfr # TSFE14-0250-062
COMPARE
213 1/8 1/4
$308.00USD / Pkg of 1
ChemFluor TSFE14-0250-047 FEP Tubing, 159 psi @ 70F, 5/32 X 1/4, 50'
Buy More Save More
EW-96001-12 Mfr # TSFE14-0250-047
COMPARE
159 5/32 1/4
$209.00USD / Pkg of 1
ChemFluor TSFE14-0250-031-50 FEP, 3/16 x 1/4" Tubing, 106psi Maximum at 70°F, 50 Ft
Buy More Save More
EW-96001-14 Mfr # TSFE14-0250-031-50
COMPARE
106 3/16 1/4
$170.00USD / Pkg of 1
ChemFluor TSFE14-0312-062 FEP tubing, 3/16 x 5/16, pack of 50'
Buy More Save More
EW-96001-16 Mfr # TSFE14-0312-062
COMPARE
170 3/16 5/16
$367.00USD / Pkg of 1
ChemFluor TSFE14-0312-031 FEP tubing, 1/4 x 5/16, pack of 50'
Buy More Save More
EW-96001-18 Mfr # TSFE14-0312-031
COMPARE
85 1/4 5/16
$201.00USD / Pkg of 1
ChemFluor TSFE14-0375-062 FEP tubing, 1/4 x 3/8, pack of 50'
Buy More Save More
EW-96001-20 Mfr # TSFE14-0375-062
COMPARE
142 1/4 3/8
$435.00USD / Pkg of 1
ChemFluor TSFE14-0375-031 FEP tubing, 5/16 x 3/8, pack of 50'
Buy More Save More
EW-96001-22 Mfr # TSFE14-0375-031
COMPARE
71 5/16 3/8
$251.00USD / Pkg of 1
ChemFluor TSFE14-0437-031 FEP tubing, 3/8 x 7/16, pack of 50'
Buy More Save More
EW-96001-26 Mfr # TSFE14-0437-031
COMPARE
61 3/8 7/16
$280.00USD / Pkg of 1
ChemFluor TSFE14-0500-062 FEP Tubing, 3/8 X 1/2, 106 psi @ 70F, 50'
Buy More Save More
EW-96001-28 Mfr # TSFE14-0500-062
COMPARE
106 3/8 1/2
$598.00USD / Pkg of 1
ChemFluor TSFE14-0500-031 FEP tubing, 7/16 x 1/2, pack of 50'
Buy More Save More
EW-96001-30 Mfr # TSFE14-0500-031
COMPARE
53 7/16 1/2
$350.00USD / Pkg of 1
ChemFluor TSFE14-0562-031 FEP tubing, 1/2 x 9/16, pack of 50'
Buy More Save More
EW-96001-34 Mfr # TSFE14-0562-031
COMPARE
47 1/2 9/16
$391.00USD / Pkg of 1
ChemFluor TSFE14-0625-062 FEP tubing, 1/2 x 5/8, pack of 50'
Buy More Save More
EW-96001-36 Mfr # TSFE14-0625-062
COMPARE
85 1/2 5/8
$802.00USD / Pkg of 1
ChemFluor FEP, 5/8 x 11/16" Tubing, 39psi Maximum at 70°F, 50 Ft
Buy More Save More
EW-96001-42 Mfr # TSFE14-0687-031
COMPARE
39 5/8 11/16
$574.00USD / Pkg of 1
ChemFluor TSFE14-0750-062 FEP tubing, 5/8 x 3/4, pack of 50'
Buy More Save More
EW-96001-44 Mfr # TSFE14-0750-062
COMPARE
71 5/8 3/4
$1,042.00USD / Pkg of 1
ChemFluor TSFE14-0875-062 FEP tubing, 3/4 x 7/8, pack of 50'
Buy More Save More
EW-96001-52 Mfr # TSFE14-0875-062
COMPARE
61 3/4 7/8
$1,444.00USD / Pkg of 1
ChemFluor TSFE14-1000-062 FEP tubing, 7/8 x 1, pack of 50'
Buy More Save More
EW-96001-56 Mfr # TSFE14-1000-062
COMPARE
53 7/8 1
$1,596.00USD / Pkg of 1
ChemFluor TSFE14-1125-062 FEP tubing, 1 x 1-1/8, pack of 50'
Buy More Save More
EW-96001-60 Mfr # TSFE14-1125-062
COMPARE
47 1 1
$1,622.00USD / Pkg of 1
Showing 1 - 20 of 21