Chemfluor PFA Tubing 18618
Applications:Pharmaceutical processes, cable covering, electrical applications.
Characteristics:Excellent temperature range, chemically inert, tough, flexible, low friction coefficient, Ozone-, UV-, and stress and crack resistant. Translucent.
Certification:FDA compliant. Meets USP Class VI requirements.
Temperature range:–320 to 500°F (–195 to 260°C).
Sterilize:With autoclave, ethylene oxide, or use dry heat.
$165.00 - $2,044.00USD / Pkg of 1
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
ChemFluor PFA tubing, 1/16 x 1/8, pack of 50'
Buy More Save More
EW-96005-02 Mfr # TSPF35-0125-031-50
COMPARE
412 1/16 1/8
$165.00USD / Pkg of 1
ChemFluor TSPF35-0187-031 PFA tubing, 1/8 x 3/16, pack of 50'
Buy More Save More
EW-96005-08 Mfr # TSPF35-0187-031
COMPARE
150 1/8 3/16
$274.00USD / Pkg of 1
ChemFluor TSPF35-0250-062 PFA tubing, 1/8 x 1/4, pack of 50'
Buy More Save More
EW-96005-10 Mfr # TSPF35-0250-062
COMPARE
225 1/8 1/4
$530.00USD / Pkg of 1
ChemFluor PFA Tubing, 5/32 X 1/4, Pack of 50'
Buy More Save More
EW-96005-12 Mfr # TSPF35-0250-047-50
COMPARE
169 5/32 1/4
$472.00USD / Pkg of 1
ChemFluor TSPF35-0250-031 PFA tubing, 3/16 x 1/4, pack of 50'
Buy More Save More
EW-96005-14 Mfr # TSPF35-0250-031
COMPARE
180 3/16 1/4
$357.00USD / Pkg of 1
ChemFluor TSPF35-0312-062 PFA tubing, 3/16 x 5/16, pack of 50'
Buy More Save More
EW-96005-16 Mfr # TSPF35-0312-062
COMPARE
144 3/16 5/16
$750.00USD / Pkg of 1
ChemFluor TSPF35-0312-031 PFA tubing, 1/4 x 5/16, pack of 50'
Buy More Save More
EW-96005-18 Mfr # TSPF35-0312-031
COMPARE
90 1/4 5/16
$468.00USD / Pkg of 1
ChemFluor TSPF35-0375-062 PFA tubing, 1/4 x 3/8, pack of 50'
Buy More Save More
EW-96005-20 Mfr # TSPF35-0375-062
COMPARE
150 1/4 3/8
$740.00USD / Pkg of 1
ChemFluor TSPF35-0437-031 PFA tubing, 3/8 x 7/16, pack of 50'
Buy More Save More
EW-96005-26 Mfr # TSPF35-0437-031
COMPARE
64 3/8 7/16
$676.00USD / Pkg of 1
ChemFluor TSPF35-0500-062 PFA tubing, 3/8 x 1/2, pack of 50'
Buy More Save More
EW-96005-28 Mfr # TSPF35-0500-062
COMPARE
113 3/8 1/2
$998.00USD / Pkg of 1
ChemFluor TSPF35-0500-031 PFA tubing, 7/16 x 1/2, pack of 50'
Buy More Save More
EW-96005-30 Mfr # TSPF35-0500-031
COMPARE
56 7/16 1/2
$748.00USD / Pkg of 1
ChemFluor TSPF35-0625-062 PFA tubing, 1/2 x 5/8, pack of 50'
Buy More Save More
EW-96005-36 Mfr # TSPF35-0625-062
COMPARE
90 1/2 5/8
$1,420.00USD / Pkg of 1
ChemFluor TSPF35-0750-062 PFA tubing, 5/8 x 3/4, pack of 50'
Buy More Save More
EW-96005-44 Mfr # TSPF35-0750-062
COMPARE
75 5/8 3/4
$1,398.00USD / Pkg of 1
ChemFluor TSPF35-0875-062 PFA tubing, 3/4 x 7/8, pack of 50'
Buy More Save More
EW-96005-52 Mfr # TSPF35-0875-062
COMPARE
64 3/4 7/8
$2,044.00USD / Pkg of 1
Showing 1- 14 of 14