Chemfluor® PTFE Tubing
Applications: Chemical transfer and pharmaceutical processes.
Characteristics: Most flexible fluoropolymer. Excellent chemical compatibility. Translucent.
Certification: FDA compliant.
Temperature range: –450 to 550°F (–268 to 288°C).
Sterilize: Autoclave, ethylene oxide, or dry heat.
$127.00 - $1,810.00USD / Pkg of 1
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
ChemFluor TSPTFE-0125-031 PTFE Tubing, 1/16" x 1/8"; 50 ft/Pk
Buy More Save More
EW-96000-02 Mfr # TSPTFE-0125-031
COMPARE
156 1/16 1/8
$127.00USD / Pkg of 1
ChemFluor TSPTFE-0250-062-50 PTFE Tubing, 1/8" x 1/4"; 50 ft/Pk
Buy More Save More
EW-96000-04 Mfr # TSPTFE-0250-062-50
COMPARE
156 1/8 1/4
$301.00USD / Pkg of 1
ChemFluor TSPTFE-0250-047-50 PTFE Tubing, 5/32" x 1/4"; 50 ft/Pk
Buy More Save More
EW-96000-06 Mfr # TSPTFE-0250-047-50
COMPARE
117 5/32 1/4
$290.00USD / Pkg of 1
ChemFluor TSPTFE-0250-031-50 PTFE Tubing, 3/16" x 1/4"; 50 ft/Pk
Buy More Save More
EW-96000-08 Mfr # TSPTFE-0250-031-50
COMPARE
78 3/16 1/4
$232.00USD / Pkg of 1
ChemFluor TSPTFE-0312-062-50 PTFE Tubing, 3/16" x 5/16"; 50 ft/Pk
Buy More Save More
EW-96000-10 Mfr # TSPTFE-0312-062-50
COMPARE
125 3/16 5/16
$442.00USD / Pkg of 1
ChemFluor TSPTFE-0312-031 PTFE Tubing, 1/4" x 5/16"; 50 ft/Pk
Buy More Save More
EW-96000-12 Mfr # TSPTFE-0312-031
COMPARE
63 1/4 5/16
$224.00USD / Pkg of 1
ChemFluor TSPTFE-0375-062-50 PTFE Tubing, 1/4" x 3/8"; 50 ft/Pk
Buy More Save More
EW-96000-14 Mfr # TSPTFE-0375-062-50
COMPARE
104 1/4 3/8
$640.00USD / Pkg of 1
ChemFluor TSPTFE-0375-031-50 PTFE Tubing, 5/16" x 3/8"; 50 ft/Pk
Buy More Save More
EW-96000-16 Mfr # TSPTFE-0375-031-50
COMPARE
52 5/16 3/8
$307.00USD / Pkg of 1
ChemFluor TSPTFE-0500-062 PTFE Tubing, 3/8" x 1/2"; 50 ft/Pk
Buy More Save More
EW-96000-18 Mfr # TSPTFE-0500-062
COMPARE
78 3/8 1/2
$936.00USD / Pkg of 1
ChemFluor TSPTFE-0500-031 PTFE Tubing, 7/16" x 1/2"; 50 ft/Pk
Buy More Save More
EW-96000-20 Mfr # TSPTFE-0500-031
COMPARE
39 7/16 1/2
$404.00USD / Pkg of 1
ChemFluor TSPTFE-0625-062 PTFE Tubing, 1/2" x 5/8"; 50 ft/Pk
Buy More Save More
EW-96000-26 Mfr # TSPTFE-0625-062
COMPARE
63 1/2 5/8
$1,274.00USD / Pkg of 1
ChemFluor TSPTFE-0625-031 PTFE Tubing, 9/16" x 5/8"; 50 ft/Pk
Buy More Save More
EW-96000-28 Mfr # TSPTFE-0625-031
COMPARE
31 9/16 5/8
$522.00USD / Pkg of 1
ChemFluor TSPTFE-0750-062 PTFE Tubing, 5/8" x 3/4"; 50 ft/Pk
Buy More Save More
EW-96000-30 Mfr # TSPTFE-0750-062
COMPARE
52 5/8 3/4
$1,810.00USD / Pkg of 1
Showing 1- 13 of 13