This product is now discontinued, View Related Products

Acros Organics Chlorotrimethylsilane

Consistent, high quality organics and fine chemicals at great savings
The Acros Organics brand delivers consistent, high-quality organics and fine chemicals at great savings. It also focuses on the evolving trends in organic chemistry, supporting research, development and product activities through a range of off-the-shelf products and expertise in sourcing, manufacturing and technology partnerships. Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Call For Price

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
CAS Number Volume (mL)
Availability Pricing
Acros Organics AC110120010 Chlorotrimethylsilane 98% (1l) EW-88100-50 Mfr # AC110120010
COMPARE
75-77-4 1000
Call For Price
Acros Organics AC110120250 Chlorotrimethylsilane 98% (25ml) EW-88100-51 Mfr # AC110120250
COMPARE
75-77-4 25
Call For Price
Acros Organics AC110121000 Chlorotrimethylsilane 98% (100ml) EW-88100-52 Mfr # AC110121000
COMPARE
75-77-4 100
Call For Price
Acros Organics AC110122500 Chlorotrimethylsilane 98% (250ml) EW-88100-53 Mfr # AC110122500
COMPARE
75-77-4 250
Call For Price
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat