Chromatography/Analytical Gas Oxygen Traps
Reduce residual oxygen content to less than 0.1 ppm with standard gas chromatography flow rates. Indicating material changes color from bright green to gray when depleted. Plastic sleeve protects glass housing in case of breakage. Traps measure 10-1/4"L x 1-3/8" dia. Shpg wt 1 lb (0.5 kg).
$186.00 - $203.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Air Pressure (psig) Max Flow Rate (mL/min) Purity
Availability Pricing
COMPARE
$186.00USD / Each
COMPARE
$203.00USD / Each