Cold Water Housing with Transparent, Styrene Acrylonitrile (SAN) Sump
Model 01509-02 available with pressure relief valve
  • Constructed of durable Styrene Acrylonitrile (SAN)
Cold Water Housing with Transparent, Styrene Acrylonitrile (SAN) Sump. Talc-filled polypropylene cap. Maximum temperature is 125°>F (52°>C); maximum pressure is 125 psi. Accepts all DOE cartridges. Buna N seal. Model 01509-02 includes a pressure relief valve.
$52.50 - $622.75USD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cartridge Length (in) Flow Rate Housing Material
Availability Pricing
COMPARE
10 1 to 10 GPM Talc-Filled Polypropylene
$52.50USD / Each
COMPARE
10 1 to 10 GPM Pure Polypropylene
$132.75USD / Each
COMPARE
20 1 to 15 GPM Pure Polypropylene
$110.50USD / Each
COMPARE
10 1 to 10 GPM Glass-Reinforced Nylon
$214.00USD / Each
COMPARE
20 1 to 15 GPM Talc-Filled Polypropylene
$75.25USD / Each
COMPARE
10 1 to 10 GPM 304 Stainless Steel
$352.50USD / Each
COMPARE
20 1 to 15 GPM 304 Stainless Steel
$622.75USD / Each
COMPARE
10 1 to 10 GPM Styrene Acrylonitrile
$62.50USD / Each
COMPARE
10 1 to 10 GPM Styrene Acrylonitrile
$64.00USD / Each
Showing 1- 9 of 9