This product is now discontinued, View Related Products

Cole-Parmer® 1-Octanol

Consistent, high quality organics and fine chemicals at great savings
  • Quality and consistency your lab can count on
The Acros Organics brand delivers consistent, high-quality organics and fine chemicals at great savings. It also focuses on the evolving trends in organic chemistry, supporting research, development and product activities through a range of off-the-shelf products and expertise in sourcing, manufacturing and technology partnerships. Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Call For Price

6 variations of this product are available.

See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
CAS Number Volume (mL) Description
Availability Pricing
Fisher Chemical A402500 1-Octanol (Certified) (500 mL) CAS 111-87-5 EW-88018-02 Mfr # A402500
COMPARE
111875 500 1-Octanol (Certified) (500 mL) CAS 111-87-5
Call For Price
Acros Organics 1-Octanol 98% (1l) CAS 111-87-5 - Liquid EW-88152-10 Mfr # AC150630010
COMPARE
111-87-5 1000 1-Octanol 98% (1l) CAS 111-87-5 - Liquid
Call For Price
Acros Organics AC150630025 1-Octanol, 98%; 2.5 L EW-88152-11 Mfr # AC150630025
COMPARE
111-87-5 2500 1-Octanol 98% (2.5l) CAS 111-87-5 - Liquid
Call For Price
Acros Organics AC220180025 1-Octanol 99+% (2.5l) EW-88235-50 Mfr # AC220180025
COMPARE
111-87-5 1-Octanol 99+% (2.5l)
Call For Price
Acros Organics AC220185000 1-Octanol 99+% (500 mL) EW-88235-51 Mfr # AC220185000
COMPARE
111-87-5 1-Octanol 99+% (500 mL)
Call For Price
COMPARE
111-87-5 1000 1-Octanol; 1 L
Call For Price
Showing 1- 6 of 6

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat