Cole-Parmer 3-D Rocking Shakers
Gentle gyratory motion for low-foaming agitation
  • Adjust the angle of tilt to optimize mixing
  • Built-in timer and digital speed LED display ensure repeatable results
  • Triple the capacity with optional tier system
This 3-D rocking shaker performs a gentle swirling, making it ideal for delicate cell culturing, histological preparations, and DNA extractions. Choose from two sizes: small for individual use or for tight spaces, and large for larger laboratories and high-throughput applications. Platform lip and nonslip mat hold flat-sided vessels secure during mixing. Optional magnetic tier system increases capacity and can be fitted in seconds without the need for tools.
$988.25 - $1,418.50USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Load (lbs) Power (VAC)
Availability Pricing
COMPARE
6.6 120
$988.25USD / Each
COMPARE
6.6 230
$1,039.00USD / Each
COMPARE
22 120
$1,026.00USD / Each
COMPARE
22 230
$1,418.50USD / Each