Cole-Parmer 760 Torr Absolute Digital Reference Gauges, 120 VAC
Replace mercury manometers for monitoring vacuum systems
  • ±0.25% test gauge accuracy
  • 316 stainless steel wetted parts
  • Fast response—3 readings per second
Replace toxic mercury manomaters with these vacuum gauges! Use for monitoring laboratory or industrial vacuum manifolds, pumps, and systems. Large display provides better readability than mercury or liquid manometers. The 760 torr absolute scale measures atmospheric pressure when gauge port is open and reads zero at full vacuum.
Rugged epoxy-coated extruded aluminum case and rear cover, ABS/polycarbonate front bezel, front on/off push button, front accessible calibration adjustments.
$432.50 - $465.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Accuracy Resolution Description
Availability Pricing
COMPARE
±0.25% full-scale ±1 digit 1 torr Digital Gauge, 760 Torr; 1/4\"; NPT(M)
$432.50USD / Each
COMPARE
±0.25% full-scale ±1 digit 1 torr Digital Gauge, 760 torr; 1/4\" NPT(M) w / Backlight
$465.00USD / Each