This product is now discontinued, View Related Products
Cole-Parmer Benchtop Analog Gear Pump Drive Systems
Analog drives provide continuous-duty operation with Micropump® A-mount pump heads. Gradual, one-second acceleration offers smooth control of drive speed and flow rate.
The permanent-magnet, direct-current (PMDC) drive is built to last — excellent for long-term applications.
Service kits 07144-42, 74012-02, -42, and -52 include: gears, suction shoe, shaft seals, and
springs.
Service kits 74012-12, -22, and -32 include: gears, bushings, and seals.
Call For Price
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Flow Rate (mL/rev) Max Flow Rate (mL/min) Power (VAC)
Availability Pricing
COMPARE
0.092 331.2 115
Call For Price
COMPARE
0.092 331.2 220
Call For Price
COMPARE
0.316 1140 115
Call For Price
COMPARE
0.316 1140 220
Call For Price
COMPARE
0.64 2300 115
Call For Price
COMPARE
0.64 2300 220
Call For Price
COMPARE
0.91 3280 115
Call For Price
COMPARE
0.91 3280 220
Call For Price
COMPARE
1.17 4210 115
Call For Price
COMPARE
1.17 4210 220
Call For Price
COMPARE
0.261 939.6 115
Call For Price
COMPARE
0.261 939.6 220
Call For Price
COMPARE
0.092 331.2 115
Call For Price
COMPARE
0.092 331.2 220
Call For Price
COMPARE
0.017 85 115
Call For Price
COMPARE
0.017 61.2 220
Call For Price
Showing 1- 16 of 16