Cole-Parmer® Essentials Burettes (Burets)
Class B Burette (Burets) meets your basic measuring and dispensing requirements!
Manufactured from high quality borosilicate glass, burettes (Burets) are designed for accuracy and durability. Tips have been carefully drawn for reliable drainage rate. Burettes feature non-contaminating 2mm straight bore PTFE stopcock, and red colored permanent graduations.
$108.00 - $147.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Tolerance
Availability Pricing
COMPARE
10 0.04
$108.00USD / Each
COMPARE
25 0.1
$116.00USD / Each
COMPARE
50 0.1
$126.00USD / Each
COMPARE
100 0.2
$147.00USD / Each