Cole-Parmer® Cable Zip Ties, White Nylon
Won’t corrode or stretch
  • Holds securely—even in vibrating, oily, or wet environments
  • Withstands temperatures to 85 °C (185 °F)
  • Available in a variety of tensile strengths from 11 to 175 pounds
Injection-molded of high-quality 6/6 nylon, these solid cable/zip ties provide a lasting solution for organization of your wire and cable bundles. They have a durable locking mechanism that holds securely, even when used to bundle vibrating or flexing materials. They will not corrode or stretch and will continue to hold tension even in oily spray or wet environments. A bent tail allows for easy pickup of the tie and threading through the lock.
Able to withstand temperatures to 85 °C (185 °F), these cable/zip ties have a melting point of 257 °C (495 °F) and remain flexible to -40 °C (-40 °F). Efficient range of ties may be higher or lower in your specific application. They are RoHS compliant, UL recognized, and UL 94 V-2 flammability rated.
Easily install by wrapping the tie around the bundle and threading the tail through the locking mechanism. Pull tight by hand and cut the tail to complete or use a tensioning tool—available separately. For mounting wires and cables to walls or other surfaces, use Cole-Parmer® Self Adhering Cable Zip Tie Mounts—available separately.
ITEM(S) ON SALE
$1.50 - $93.00USD / Bag of 1000
25 variations of this product are available.
See All Product Options Below
25 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Bundle Diameter (in) Length (in) Tensile Strength (lbs)
Availability Pricing
COMPARE
2 1/10 7 29/32 40
$57.00USD / Bag of 1000
COMPARE
4 14 19/32 120
$19.20USD / Bag of 50
COMPARE
7 19/64 28 2/5 120
$38.50USD / Bag of 50
COMPARE
9 1/10 32 19/64 175
$38.50USD / Bag of 50
COMPARE
2 1/10 7 29/32 50
$10.70USD / Pkg of 100
COMPARE
3 12 50
$93.00USD / Pkg of 1000
COMPARE
29/32 3 27/32 18
$30.00USD / Bag of 1000
COMPARE
1 2/5 6 40
$34.50USD / Bag of 1000
$46.50USD / Bag of 1000
COMPARE
2 1/10 7 29/32 50
$63.50USD / Bag of 1000
COMPARE
4 14 19/32 50
$21.50USD / Bag of 100
COMPARE
29/32 4 18
$6.00USD / Pkg of 100
COMPARE
4 14 19/32 120
$28.50USD / Pkg of 100
COMPARE
27/64 2 1/2 11
$1.50USD / Case of 100
COMPARE
1 3/8 6 18
$3.50USD / Case of 100
COMPARE
2 5/64 8 18
$4.50USD / Case of 100
COMPARE
7/8 4 40
$3.50USD / Case of 100
COMPARE
1 3/8 6 40
$6.00USD / Case of 100
COMPARE
2 5/64 8 40
$10.30USD / Case of 100
COMPARE
2 63/64 12 40
$10.50USD / Case of 100
COMPARE
4 1/50 14 1/2 40
$16.00USD / Case of 100
Showing 1 - 20 of 25