Cole-Parmer Canned Motor Bronze Standard Centrifugal Pumps
Transfer water using as little as 12 watts
  • Noryl® body models are available
  • Whisper quiet - less than 40 db noise level
  • Magnetically driven means no mechanical seal to potentially fail
  • Universal voltage of 100 to 240 VAC, 50/60 Hz
$256.20 - $395.50USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Inlet Connection Max Flow Rate (GPM) Max Head (ft)
Availability Pricing
COMPARE
0.5" sweat 3.5 6
$256.20USD / Each
COMPARE
0.5" union 3.5 6
$336.60USD / Each
COMPARE
5.5 11
$377.20USD / Each
COMPARE
0.5" union 5.5 11
$336.60USD / Each
COMPARE
0.5" sweat 5.5 11
$395.50USD / Each