Cole-Parmer Centrifuge Tube Vacuum Filtration System
Pre-sterilized filtration systems for every application
  • Gamma sterilized to reduce the risk of contamination
  • Made with a low protein binding and surfactant-free PES membrane
  • Available in three membrane pore size options for an assortment of applications
The centrifuge tube filtration systems include the upper funnel and a 15- or 50-mL centrifuge tube. This allows for filtration directly into the centrifuge tube, reducing the risk of spills by eliminating unnecessary fluid transfer. These capacities are available in in 0.1 um, 0.2 um, and 0.45 pore sizes.
These systems can be used in conjunction with the optional pedestal or cradle ring to create a hands-free filtration system.
$190.00 - $248.00USD / Case of 24
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Membrane Diameter (mm) Pore Size (µm)
Availability Pricing
COMPARE
50 40 0.2
$190.00USD / Case of 24
COMPARE
50 40 0.45
$190.00USD / Case of 24
COMPARE
15 40 0.2
$190.00USD / Case of 24
COMPARE
15 40 0.45
$190.00USD / Case of 24
COMPARE
15 40 0.1
$248.00USD / Case of 24
COMPARE
50 40 0.1
$224.00USD / Case of 24
Showing 1- 6 of 6
Need Help with this product?
Call Us At1-847-549-7600
Live chat