Cole-Parmer Certified VOA Vials
Borosilicate glass vials meet EPA guidelines for contamination-free containers
  • Choose between 20-, 40-, and 60-mL capacities
  • Select standard screw-top or open-top septa caps
  • Screw-top caps feature either PTFE or septa liner
Level 1 VOA vials meet EPA recommended guidelines for contamination-free sample containers and include a Certificate of Analysis. Septa offered as a liner option for closed caps to ensure greater seal integrity.
$76.00 - $162.00USD / Case of 72
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Description Tube Color Volume (mL)
Availability Pricing
COMPARE
Clear VOA Vials, 40 mL, PTFE-Lined Caps, Economy Case; 144/Cs Clear 40
$101.00USD / Case of 144
COMPARE
Amber VOA Vials, 40 mL, PTFE-Lined Caps, Economy Case; 144/Cs Amber 40
$109.00USD / Case of 144
COMPARE
Amber VOA Vials, 40 mL, Open-Top Green Septa Caps, Economy Case; 144/Cs Amber 40
$154.00USD / Case of 144
COMPARE
Clear VOA Vials, 40 mL, Open-Top Septa Caps; 144/Cs Clear 40
$119.00USD / Case of 144
COMPARE
Clear VOA Vials, 40 mL, Open-Top Green Septa Caps, Economy Case; 144/Cs Clear 40
$119.00USD / Case of 72
COMPARE
Clear VOA Vials, 40 mL, Open-Top Septa Caps; 72/Cs Clear 40
$76.00USD / Case of 72
COMPARE
Amber VOA Vials, 60 mL, Open-Top Green Septa Caps; 72/Cs Amber 60
$91.50USD / Case of 72
COMPARE
Amber VOA Vials, 40 mL, Open-Top Green Septa Caps; 144/Cs Amber 40
$157.00USD / Case of 72
COMPARE
Clear VOA Vials, 20 mL, PTFE-Lined Caps; 144/Cs Clear 20
$115.00USD / Case of 72
COMPARE
Amber VOA Vials, 20 mL, Open-Top Septa Caps; 144/Cs Amber 20
$162.00USD / Case of 144
Showing 1- 10 of 10
Need Help with this product?
Call Us At1-847-549-7600
Live chat