Cole-Parmer Chemical Feed Systems
Complete and preassembled chemical feed system where 24/7 use is required
  • Shipped complete - "plug and play" installation is as simple as providing power and fluid connections
  • Pump(s) mounted on a PVC based skid—choose from multiple configurations
  • Preconfigured systems are designed for use in either pH or ORP applications
Systems will include metering pumps from Walchem. Other manufacturers such as PulsaFeeder or LMI can easily be substituted by calling one of our Application Specialists.
The skid is available in either single- or dual-pump, configurations and is preplumbed for easy installation. pH/ORP systems inject acid or base into your fluid system to maintain a desired set point—but will require a separate controller, or connect to your existing controller.
If a more accurate and reliable measurement system is needed, we offer ORP controller based systems that can also adjust the measured fluid by adding acid and/or base. Controller included. All you have to do is to hook up the fluid and power connections.
Although these preconfigured systems are designed for use in either pH or ORP applications, any number of custom configurations are achievable! These redesigns can be accomplished with a simple phone call to our Application Specialists to ensure that all of your system needs are met.
$9,140.00 - $12,600.00USD / Each
32 variations of this product are available.
See All Product Options Below
32 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (GPH) Max Pressure (PSI) Power (VAC)
Availability Pricing
COMPARE
7.5 7.2 115
$10,000.00USD / Each
COMPARE
16.2 3.4 115
$10,200.00USD / Each
COMPARE
23.8 2.06 115
$10,200.00USD / Each
COMPARE
2.2 10.3 230
$9,150.00USD / Each
COMPARE
7.5 7.2 230
$10,000.00USD / Each
COMPARE
16.2 3.4 230
$10,200.00USD / Each
COMPARE
23.8 2.06 230
$10,200.00USD / Each
COMPARE
16.2 3.4 115
$12,100.00USD / Each
COMPARE
7.5 7.2 230
$11,900.00USD / Each
COMPARE
16.2 3.4 230
$12,100.00USD / Each
COMPARE
23.8 2.06 230
$12,600.00USD / Each
COMPARE
2.2 10.3 115
$9,140.00USD / Each
COMPARE
7.5 7.2 115
$9,990.00USD / Each
COMPARE
2.2 10.3 115
$10,100.00USD / Each
COMPARE
16.2 3.4 115
$12,000.00USD / Each
COMPARE
23.8 2.06 115
$12,600.00USD / Each
COMPARE
2.2 10.3 230
$10,100.00USD / Each
COMPARE
7.5 7.2 230
$11,900.00USD / Each
COMPARE
16.2 3.4 230
$12,100.00USD / Each
COMPARE
23.8 2.06 230
$12,600.00USD / Each
Showing 1 - 20 of 32
Need Help with this product?
Call Us At1-847-549-7600
Live chat