Cole-Parmer Chemically Inert Metering Valves
Excellent for metering corrosive fluids and gases — all wetted parts are CTFE and PTFE. Eight turns will fully open or close valves. Maximum pressure is 75 psi and maximum temperature is 150°F (93°C); orifice size is 1/4".
$82.00 - $103.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate For Air (mL/min) Max Flow Rate For Water (mL/min) Port Size (NPT(F))
Availability Pricing
COMPARE
600 36 0.125
$103.00USD / Each
COMPARE
3000 180 0.125
$103.00USD / Each
COMPARE
30000 1800 0.125
$103.00USD / Each
COMPARE
300000 9000 0.375
$82.00USD / Each