This product is now discontinued, View Related Products
Cole-Parmer® Class A Griffin Low-Form Graduated Glass Beakers
Double-scale white graduations for easy reading up and down!
  • Double-scale graduations for easy measuring while pouring in or out
  • Autoclavable
These low-form beakers are constructed of Class A borosilicate glass to meet ASTM E960 specifications. Beakers can withstand temperatures up to 752°F (400°C) for short-term exposure and 445°F (230°C) for long-term exposure.
Call For Price
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
COMPARE
10 Borosilicate Glass
Call For Price
COMPARE
25 5 Borosilicate Glass
Call For Price
COMPARE
50 10 Borosilicate Glass
Call For Price
COMPARE
100 10 Borosilicate Glass
Call For Price
COMPARE
150 20 Borosilicate Glass
Call For Price
COMPARE
250 25 Borosilicate Glass
Call For Price
COMPARE
400 25 Borosilicate Glass
Call For Price
COMPARE
600 50 Borosilicate Glass
Call For Price
COMPARE
1000 50 Borosilicate Glass
Call For Price
COMPARE
2000 100 Borosilicate Glass
Call For Price
COMPARE
3000 125 Borosilicate Glass
Call For Price
COMPARE
5000 250 Borosilicate Glass
Call For Price
Showing 1- 12 of 12