This product is now discontinued, View Related Products
Cole-Parmer Class I Ductless Workstations
Portable, compact workstations are completely self-contained
  • Integral recessed work surface contains spills
  • Top airflow is 100 CFM and has high, medium and low settings
Compact benchtop workstations are ideal for your general chemistry, tissue research, forensic applications, and academic demonstrations. The prefilter and activated carbon filter remove fumes, odors, and nonhazardous chemicals.
Clear acrylic enclosure lets you easily view workstation contents from three sides. Front cutaways allow ergonomic use.
Larger workstations 33730-26 and -28 have a hinged sash that enables you to have a completely open front.
Filters for both size workstations are easily replaced by removing the prefilter and then unclipping the carbon filter from the rear baffle. Workstations 33730-26 and -28 feature a filter change light, but it is recommended filters be changed every 60 to 90 days with normal use. Models 33730-22 and -24 have an electrical port exit on the left side. Models 33730-26 and -28 have electrical port exits on both sides of workstation.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Exterior Height (in) Exterior Width (in) Power (VAC)
Availability Pricing
COMPARE
14.5 12 115
Call For Price
COMPARE
14.5 12 230
Call For Price
COMPARE
20 18 115
Call For Price
COMPARE
20 18 230