Cole-Parmer® Cleanroom Hose Barb Fittings, Y Union, Natural Kynar®
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Quick and easy assembly without tools
  • Single barb design tightly “grips” the tubing
  • Biopharma compatible and USP 661 and BPOG compliant
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Easily combine or split fluid flow using Y unions. A single barb on each end of the fitting creates a tight seal to keep the tubing in place. An anti-rotation device on each port prevents tubing from rotating, minimizing kinks. These natural Kynar® fittings provide excellent chemical, flame, gamma radiation, UV, and weather resistance. They also exhibit good mechanical properties, low moisture absorption, and good machinability.
$26.00 - $230.00USD / Pkg of 10
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Barb Size / Tubing Id (Port 3) (in)
Availability Pricing
COMPARE
3/32 3/32 3/32
$26.00USD / Pkg of 10
COMPARE
5/8 5/8 5/8
$169.00USD / Pkg of 10
COMPARE
3/4 3/4 3/4
$210.00USD / Pkg of 10
COMPARE
1 1 1
$230.00USD / Pkg of 10
COMPARE
1/16 1/16 1/16
$26.00USD / Pkg of 10
COMPARE
5/32 5/32 5/32
$40.00USD / Pkg of 10
COMPARE
1/8 1/8 1/8
$31.50USD / Pkg of 10
COMPARE
3/16 3/16 3/16
$40.00USD / Pkg of 10
COMPARE
1/4 1/4 1/4
$46.25USD / Pkg of 10
COMPARE
5/16 5/16 5/16
$113.50USD / Pkg of 10
COMPARE
3/8 3/8 3/8
$114.00USD / Pkg of 10
COMPARE
1/2 1/2 1/2
$128.00USD / Pkg of 10
Showing 1- 12 of 12