Cole-Parmer Conical-Bottom Centrifuge Tubes, PP
Separate samples without fear of leakage
  • Flat polyethylene tube cap with compressible liner creates stable seal
  • Bold black graduations are easy-to-read and accurate to ±2%
  • Large white marking area simplifies identification
Clear polypropylene (PP) conical bottom centrifuge tubes withstand temperatures from -80 to 120°C (-112 to 248°F). The 15-mL tubes have a maximum RCF range of 12,000 g while the 50-mL tubes can perform at speeds up to 16,000 RCF x g. Tubes are available sterile-packed in resealable bags or recyclable polypropylene racks.
$91.35 - $133.30USD / Case of 300
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tube Capacity (mL) Description
Availability Pricing
COMPARE
15 Centrifuge Tube, 15 mL, Resealable Bag; 500/cs
$112.30USD / Case of 500
COMPARE
15 Centrifuge Tube, 15 mL, Plastic Rack; 500/cs
$91.35USD / Case of 300
COMPARE
50 Centrifuge Tube, 50 mL, Resealable Bag; 500/cs
$133.30USD / Case of 500
COMPARE
50 Centrifuge Tube, 50 mL, Platsic Rack; 500/cs
$106.60USD / Case of 300