Cole-Parmer Controlled Wall Mount Panel for pH or ORP Control Applications
Pump-less systems are pre-fabricated and tested in an easy-to-use wall mount system
The controller allows for a user friendly set up where the system water is routed through a highly reliable mechanical flow switch to assure chemical feed is limited to periods of sampling and flow.
The water then enters a see through flow cell which houses the sensor and a convenient sample tap.
The micro-controller based unit accepts one probe input, either pH or ORP and, in turn, controls the onboard chemical feed pump.
Treatment chemical is fed on-board to the discharge of the flow cell where it returns to
the recirculating water. All this is done in a small space.
Two different systems are preconfigured and built depending on your application - pH or ORP.
Any brand of metering pump can be selected to work with these systems.
$3,550.00 - $3,690.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Wall Mount pH Control Panel System, with flow cell, switch, and sensor
$3,550.00USD / Each
COMPARE
Wall Mount ORP Control Panel System, with flow cell, switch, and sensor
$3,690.00USD / Each
Showing 1- 2 of 2
Need Help with this product?
Call Us At1-847-549-7600
Live chat