Cole-Parmer® Correlated Flowmeters with Valves
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. Valved meters have valves mounted at the bottom/inlet, typically used for positive pressure with liquids and gases. The valve can be reconfigured to mount at the top/outlet, often for vacuum applications. All correlated flowmeters come with data sheets for water and air at standard temperature and pressure (STP).
$189.00 - $389.00USD / Each
63 variations of this product are available.
See All Product Options Below
63 PRODUCT OPTIONS
Compareitem specifications
Material Valve Max Flow Rate (mL/min)
Availability Pricing
COMPARE
Aluminum 18.7
$189.00USD / Each
COMPARE
Aluminum 49.1
$195.00USD / Each
COMPARE
Aluminum 137
$195.00USD / Each
COMPARE
Aluminum 264
$195.00USD / Each
COMPARE
Aluminum 1682
$195.00USD / Each
COMPARE
Aluminum 8678
$195.00USD / Each
COMPARE
Aluminum 16737
$195.00USD / Each
COMPARE
PTFE and Kel-F® 92
$373.00USD / Each
COMPARE
PTFE and Kel-F® 1093
$373.00USD / Each
COMPARE
316 SS 92
$323.00USD / Each
COMPARE
316 SS 2313
$323.00USD / Each
COMPARE
316 SS 3807
$323.00USD / Each
COMPARE
Chrome-plated brass 49.1
$219.00USD / Each
COMPARE
Chrome-plated brass 92
$219.00USD / Each
COMPARE
Chrome-plated brass 264
$219.00USD / Each
COMPARE
Chrome-plated brass 374
$219.00USD / Each
COMPARE
Chrome-plated brass 814
$219.00USD / Each
COMPARE
Chrome-plated brass 844
$219.00USD / Each
COMPARE
Chrome-plated brass 1682
$219.00USD / Each
COMPARE
Chrome-plated brass 4562
$219.00USD / Each
Showing 1 - 20 of 63