Cole-Parmer® Deep-Well Microplates for Homogenizers
Durable polypropylene microplates resist perforation by grinding media
  • Alphanumeric marks for well identification
  • Use microplates with sealing mats to prevent spills and well-to-well contamination
Ideal for use with Cole-Parmer® homogenizers, these deep-well plates serve many application purposes, such as tissue homogenization and cell lysis.
$12.00 - $1,825.00USD / Case of 100
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Number of Wells Well Volume (mL) Description
Availability Pricing
COMPARE
96 1 Deep-Well Microplate, 96 x 1 mL, Round-Bottom Wells; 100/PK
$916.00USD / Pkg of 100
COMPARE
96 1 Deep-Well Microplate, 96 x 1 mL, Round-Bottom Wells; 1/EA
$12.00USD / Each
COMPARE
96 2.4 Deep-Well Microplate, 96 x 2.4 mL, Square Wells; 1/EA
$17.00USD / Each
COMPARE
48 5 Deep-Well Microplate, 48 x 5 mL, Rectangular Wells, 1/EA
$19.00USD / Each
COMPARE
24 10 Deep-Well Microplate, 24 x 10 mL, Square Wells, 1/EA
$19.00USD / Each
COMPARE
96 2.4 Deep-Well Microplate, 96 x 2.4 mL, Square Wells; 100/PK
$916.00USD / Pkg of 100
COMPARE
48 5 Deep-Well Microplate, 48 x 5 mL, Rectangular Wells, 100/PK
$1,423.00USD / Case of 100
COMPARE
24 10 Deep-Well Microplate, 24 x 10 mL, Square Wells, 100/PK
$1,825.00USD / Case of 100
COMPARE
96 1.4 96 Square Well Reinforced Titer Plate; 50/CS
$250.00USD / Case of 50
Showing 1- 9 of 9