Cole-Parmer Desiccator Kits
Convenient kits include a desiccator, desiccator plate, and desiccant
  • Ideal working depth for your drying application
  • Polystyrene desiccant-filled discs indicate when recharge is needed
Desiccator has a chemical-resistant polypropylene body, polycarbonate cover, and neoprene O-ring for a secure seal — use grease on the flange to make the unit completely airtight. The polypropylene desiccator plate allows a proper working height for samples and desiccant placement below.
The clear polystyrene, 100-mm diameter, desiccant-filled discs fit perfectly at the bottom of the desiccator. Visually inspect whether the silica gel requires recharge — desiccant color changes from blue to pink as it absorbs moisture. Regenerate desiccant by heating discs in a 122°F (50°C) oven overnight.
$169.00 - $183.00USD / Pkg of 1
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Desiccator and Desiccant Kit; 6" Diameter
$169.00USD / Pkg of 1
COMPARE
Desiccator and Desiccant Kit; 8" Diameter
$177.00USD / Pkg of 1
COMPARE
Desiccator and Desiccant Kit; 10" Diameter
$183.00USD / Pkg of 1
Showing 1- 3 of 3
Need Help with this product?
Call Us At1-847-549-7600
Live chat