Cole-Parmer Digital Metering Pumps
Superior quality pumps at affordable prices
  • Combine engineering excellence with quality materials of construction
  • Dual output adjustment, stroke length, and cycle time
  • Feature double-ball inlet and outlet cartridge-type check valves and heavy-duty, precision molded polypropylene pump head
Digital units offer pinpoint accuracy. They feature an LED readout, 4 - 50 mA, and a 4000:1 turndown ratio. The variable units offer the same digital control with the additional variable speed control capability.
$1,090.00 - $1,580.00USD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (Strokes/min) Description
Availability Pricing
COMPARE
Digital Control Metering Pump; 6 L/hr, 115 VAC, 60 Hz
$1,560.00USD / Each
COMPARE
Digital Control Metering Pump; 11.5 L/hr, 115 VAC, 60 Hz
$1,560.00USD / Each
COMPARE
Digital Control Metering Pump; 18.9 L/hr, 115 VAC, 60 Hz
$1,560.00USD / Each
COMPARE
Digital Control Metering Pump; 30 L/hr, 220 VAC, 50 Hz
$1,580.00USD / Each
COMPARE
Digital Control Metering Pump; 48.2 L/hr, 115 VAC, 60 Hz
$1,560.00USD / Each
COMPARE
Digital Control Metering Pump; 56.8 L/hr, 1115 VAC, 60 Hz
$1,560.00USD / Each
COMPARE
Digital Control Metering Pump; 45.4 L/hr, 220 VAC, 50 Hz
$1,580.00USD / Each
COMPARE
Digital Control Metering Pump; 4.8 L/hr, 220 VAC, 50 Hz
$1,090.00USD / Each
COMPARE
Analog Control Metering Pump; 45.4 L/hr, 220 VAC, 50 Hz
$1,090.00USD / Each
Showing 1- 9 of 9
Need Help with this product?
Call Us At1-847-549-7600
Live chat