This product is now discontinued, View Related Products
Cole-Parmer Ductless Demonstration Hood
Class DH-1 hood removes fumes and odors in classroom and group observation settings
  • Hood provides a clear and unobstructive view from all four sides
Ductless demonstration hood comes with activated carbon filter for removing nontoxic chemicals, nuisance odors, and particulates. Viewing height is 24" (61 cm); hood is made of clear acrylic, including hinged sash. Sash opening is 8" (20.3 cm). Attached work surface is phenolic resin. Front panel has the light switch, blower switch, and filter service light. Easily replace filter by removing clips in ceiling of hood; order replacement filters separately.
Order mobile table 33730-94 to move demo hood from room to room. Mobile table is height adjustable from 30" to 36" (76 to 91 cm).
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Exterior Height (in) Exterior Width (in) Power (VAC)
Availability Pricing
COMPARE
30.5 35 115
Call For Price
COMPARE
30.5 35 230