Cole-Parmer Economical pH and ORP Electrodes
Durable and low-maintenance electrodes
  • Ideal for laboratory and field applications
  • Epoxy-body housing resists impact
  • Gel-filled, seladed body is low maintenance
  • Compatible with most brands of meters
Epoxy-body electrodes are a selaled, gel-filled design requiring no maintenance. Single-junction design has a Ag/AgCl reference. Dimensions are 12.5 mm diameter x 90 mm length.
ITEM(S) ON SALE
$46.80 - $69.30USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Electrode Type
Availability Pricing
COMPARE
pH
$46.80USD / Each
$52.00USD / Each
COMPARE
ORP
$69.30USD / Each
$77.00USD / Each