Cole-Parmer® EB-50 Series Compact Portable Balances
Portable balance running on 12 V DC or AA battery operation with splash-proof keys
  • Accurate readings for indoor and outdoor environments
  • Splash-proof design with a sealed keypad
  • Large 5 digits LED display for comfortable viewing
  • Convenient automatic stabilization, scale only works on a flat surface
  • Tare push button to zero scale with an empty container
  • Visual indicators for weight overload and low battery
  • Portable unit design with auto-off for energy conservation
  • Basic weighing units in ounces, grams, and pounds
  • Keep the scale’s weight precision with an easy to use external calibration mass function
  • Ensure weight accuracy with a NIST-Traceable calibration
The Cole-Parmer ED compact portable balance is a basic analytical weighing balance for applications in education, the laboratory, and outdoor work areas without electrical outlets.
The balance runs with an AC/DC adapter for 12 V DC 500 mA or 6 x AA batteries for portability. It has a built-in auto power-off after 4 minutes of inactivity to save battery life. The splash-proof keypad and display have a low-battery indicator inside the 5-digit LCD display with 15 mm tall numbers. The balance comes with auto-calibration with the push of a button and an external mass.
ITEM(S) ON SALE
$85.14 - $160.82USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Power (VAC) Readability (g)
Availability Pricing
COMPARE
300 115 / 220 0.01
$145.34USD / Each
$169.00USD / Each
COMPARE
200 115 / 220 0.1
$85.14USD / Each
$99.00USD / Each
COMPARE
600 115 / 220 0.1
$123.84USD / Each
$144.00USD / Each
COMPARE
1200 115 / 220 0.1
$134.16USD / Each
$156.00USD / Each
COMPARE
2000 115 / 220 0.1
$160.82USD / Each
$187.00USD / Each
COMPARE
2000 115 / 220 1
$86.00USD / Each
$100.00USD / Each
COMPARE
5000 115 / 220 2
$86.00USD / Each
$100.00USD / Each
Showing 1- 7 of 7