Oakton by Cole-Parmer® Epoxy-Body General-Purpose pH and ORP Electrodes

Combine durability with reliability
  • Epoxy-body housing resists impact
  • Ideal for general purpose lab and field applications
  • Compatible with most brands of pH meters
The Ag/AgCl combination reference electrodes have an impact-resistant epoxy body. Electrodes provide a fast, stable response and are ideal for prolonged readings. Gel-filled design requires virtually no maintenance. 127 mm L x 12 mm diameter with maximum 176F (80C) temperature.
ITEM(S) ON SALE
$100.00 - $227.70USD / Each

6 variations of this product are available.

See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Applications Electrode Type Description
Availability Pricing
COMPARE
ORP Single junction ORP Electrode, single junction, epoxy body
$183.00USD / Each
COMPARE
pH Single junction Epoxy-Body pH Electrode with ATC, Large Ceramic Junction, Gel-Filled; 1/EA
$227.70USD / Each
$253.00USD / Each
COMPARE
pH Single junction pH electrode, single-junction, gel, BNC connector
$100.00USD / Each
COMPARE
pH Double junction pH electrode, combination, double-junction, gel filled
$123.50USD / Each
COMPARE
ORP Single junction ORP electrode; single-junction, gel-filled, BNC connector
$187.25USD / Each
COMPARE
ORP Double junction ORP electrode; double-junction, BNC Connector
$216.50USD / Each
Showing 1- 6 of 6

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat