Cole-Parmer® Essentials Conical-Bottom Centrifuge Tubes, Sterile
Specially designed flat threads prevent leakage
  • Polypropylene tubes are autoclavable to 250°F (121°C) Bright blue graduations are easy to read Maximum RCF range of 14,000 g DNase, RNase, pyrogen, and latex free
Available with a blue HDPE leak-proof screw cap, the clear polypropylene (PP) conical bottom centrifuge tubes feature a smooth hydrophobic internal surface to minimize wall effect. Both the tubes and caps are designed with flat threads for vacuum-type, leak-free closure at a pressure of >700 mm Hg. Bold blue graduations are easy to read, and a frosted surface area makes writing easier. Autoclavable at 121°C and 15 psi for 30 minutes. Radiation sterilized.
$150.00 - $161.00USD / Case of 500
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tube Capacity (mL) Tube Length (mm) Tube Outside Diameter (OD) (mm)
Availability Pricing
COMPARE
15 117.8 16.5
$150.00USD / Case of 500
COMPARE
50 114.3 29.2
$161.00USD / Case of 500