Cole-Parmer® Essentials Multiwell Tissue Culture Plates
Alphanumeric markings provide easy identification of individual wells
  • Stacking ring offers stability when handling tall stacks of plates
  • Each plate is sterilized by gamma irradiation
  • Packaged individually
Raised rim on each well nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross-contamination. Each plate is constructed of clear polystyrene, sterilized by gamma irradiation, and packaged individually (50/sleeve).
$214.00 - $268.25USD / Case
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Number of Wells Sterile Well Shape
Availability Pricing
COMPARE
6 Yes Flat
$214.00USD / Case of 100
COMPARE
12 Yes Flat
$268.25USD / Case
COMPARE
24 Yes Flat
$224.25USD / Case
COMPARE
48 Yes Flat
$233.50USD / Case
COMPARE
96 Yes Flat
$221.00USD / Case of 100
COMPARE
6 Yes Flat
$239.50USD / Case
COMPARE
96 Yes Flat
$236.25USD / Case of 100
COMPARE
12 Yes Flat
$235.50USD / Case
COMPARE
24 Yes Flat
$240.50USD / Case of 100
COMPARE
48 Yes Flat
$240.50USD / Case
COMPARE
96 Yes Flat
$256.50USD / Case
Showing 1- 11 of 11