Cole-Parmer® Essentials Specialty Microscope Slides, Soda Lime Glass

Specially prepared slides enhance adhesion of tissue samples to slides
  • Frosted one end on one side
  • Ground edges and 45° corners reduce the chance of cutting your fingers or tearing of gloves
  • Each slide measures 76 x 26 mm and 1.0 to 1.2-mm thick
Chemically resistant white writing surface is suitable for marking with inkjet and thermal transfer printers and permanent markers.
$16.60 - $17.20USD / Pkg of 72

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Slide Type Description
Availability Pricing
COMPARE
Positive charge Microscope Slides, Positive Charged, White-Frosted, 45° Corners; 72/PK
$17.20USD / Pkg of 72
COMPARE
Polysine™ coat Microscope Slides, Polysine™ Coated, White-Frosted, 45° Corners; 72/PK
$17.20USD / Pkg of 72
COMPARE
Silane coat Microscope Slides, Silane Coated, White-Frosted, 45° Corners; 72/PK
$16.60USD / Pkg of 72
COMPARE
Hydrophilic adhesive Microscope Slides, Hydrophilic Adhesive, White-Frosted, 45° Corners; 72/PK
$17.20USD / Pkg of 72
COMPARE
Cytology adhesive Microscope Slides, Cytology Adhesive, White-Frosted, 45° Corners; 72/PK
$17.20USD / Pkg of 72
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat