Cole-Parmer® Essentials Sterile Double Bagged Sampling Spoons, Polystyrene
Double bagged for extra protection from contamination
  • Specifically designed for muti-zone cleanrooms
  • Gamma irradiation sterilized (SAL 10-6)
  • Meets FDA guidelines for food, drug and cosmetic sampling
  • Individually sealed easy-to-open bags all packaged within an outer bag
  • Long, curved handle for improved control
Designed for use in multi-zone clean rooms, these single-use, lab sampling spoons are individually bagged then packaged within another bag for extra contamination protection. The long, ergonomic handle makes them ideal for gathering small amounts of sample material while minimizing the potential of personal contact. They reach easily into jars, bottles, and other confined areas. The curved handle end makes it easier to withdraw the spoon from containers or grab the spoon from a flat surface.
These laboratory spoons are made of FDA-grade polystyrene; gamma irradiation sterilized (SAL 10-6); free of BSE/TSE, heavy metals, BPA and phthalates; and lot traced. A Certificate of Irradiation is available upon request. Specified volumes are approximate and not intended for measuring specific volumes.
$100.00 - $105.00USD / Case of 50
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Overall Length (cm) Width (cm)
Availability Pricing
COMPARE
14.79 18.92 1.89
$105.00USD / Case of 50
COMPARE
4.93 17.14 1.42
$105.00USD / Case of 50
COMPARE
2.46 16.55 1.25
$100.00USD / Case of 50
COMPARE
1.25 15.85 1.25
$100.00USD / Case of 50