Cole-Parmer® Essentials Straight-Sided Jars, Amber Glass
Don't strain to get samples
  • Get easy access to contents with wide-mouth design
  • Store light-sensitive items safely in amber jars
Manufactured of Type III soda-lime glass, these wide mouth jars allow easy access for adding and removal of contents. Ideal for general lab use; amber jars are ideal for light sensitive samples. Pulp/vinyl lined screw caps protect content.
$30.50 - $73.50USD / Case of 24
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size Material
Availability Pricing
COMPARE
125 58-400 Glass
$72.50USD / Case of 24
COMPARE
250 70-400 Glass
$61.50USD / Case of 12
COMPARE
60 53-400 Glass
$30.50USD / Case of 24
COMPARE
500 89-400 Glass
$73.50USD / Case of 12