Cole-Parmer® Essentials Viscous Sample Collectors
Sample liquids with up to 100,000 mPas viscosity
  • Quickly draw fluids through the large bore hole—3/8" (10 mm)
  • Fits through container sampling and filling ports—13/16" (21 mm) overall diameter
  • Easily pull up thick liquids—features ergonomic, oversized draw handle
  • HDPE sampler is individually packaged and clean room manufactured
  • Choose from sterile or non-sterile, 100 or 200 mL samplers
$234.25 - $365.75USD / Pkg of 20
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (cm) Sterile Volume (mL)
Availability Pricing
COMPARE
50 No 100
$234.25USD / Pkg of 20
COMPARE
50 Yes 100
$277.75USD / Pkg of 20
COMPARE
100 No 200
$292.25USD / Pkg of 20
COMPARE
100 Yes 200
$365.75USD / Pkg of 20