Cole-Parmer® FS-3400 and VS-4000 Centrifuges
Fixed or variable speed for 6 or 8 place rotors
Centrifuges include a fixed angle rotor to spin tubes up to 10 mL. Choose from fixed or variable speed models; both models feature a 30-minute timer, suction cup rubber feet which firmly anchor them to the benchtop, and a stainless steel guard bowl to protect against spills. A safety lid latch cuts power to the motor if the lid is raised. The exterior is made of chemically resistant plastic.
What’s included: 6 place rotor, tube shield, 6-ft power cord.
$13.60 - $754.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Power (VAC)
Availability Pricing
COMPARE
3400 115
$519.25USD / Each
COMPARE
4000 115
$720.50USD / Each
COMPARE
$13.60USD / Each
COMPARE
4000 220
$754.00USD / Each