Cole-Parmer Gilmont Correlated Flowmeters with PTFE Fittings
  • Glass float also included: when glass float is used flow ranges are approximately half the values listed (for 316SS)
Correlated Flowmeters
Extremely accurate—±2% of reading or ±1 scale division, whichever is greater. Flowmeters with micro-sized tubes (models 03210-00, 03210-20, and 03234-50) are accurate to ±5% of reading or ±2% of scale whichever is greater. Computer calibrated flow charts are included for floats used with both air and water at STP. An “R factor” chart and formulas are included to convert scale readings for other gases or liquids or for floats other than glass. Order our flow analysis software below to generate flow charts specific to your individual applications.
Shielded models accept 3/8" ID tubing (sizes 4 and 5 accept 5/8" ID tubing). PTFE coupling adapters for unshielded models are not included—order separately.
$267.00 - $1,496.00USD / Each
21 variations of this product are available.
See All Product Options Below
21 PRODUCT OPTIONS
Compareitem specifications
Fluid Type Max Flow Rate (mL/min) Description
Availability Pricing
COMPARE
Water 8.6 Unshielded Correlated Flowmeter with Valve, Size 1 standard
$338.00USD / Each
COMPARE
Water 8.6 Shielded Correlated Flowmeter, Tube Size 1
$445.00USD / Each
COMPARE
Water 2.3 Shielded Correlated Flowmeter, Tube Size 0
$484.00USD / Each
COMPARE
Water 77 Unshielded Correlated Flowmeter, Tube Size 2
$363.00USD / Each
COMPARE
Water 77 Shielded Correlated Flowmeter, Tube Size 2
$452.00USD / Each
COMPARE
Water 640 Unshielded Correlated Flowmeter, Tube Size 3
$357.00USD / Each
COMPARE
Water 640 Shielded Correlated Flowmeter, Tube Size 3
$473.00USD / Each
COMPARE
Water 1820 Unshielded Correlated Flowmeter, Tube Size 4
$388.00USD / Each
COMPARE
Water 1820 Shielded Correlated Flowmeter, Tube Size 4
$572.00USD / Each
COMPARE
Water 4100 Unshielded Correlated Flowmeter, Tube Size 5
$417.00USD / Each
COMPARE
Water 4100 Shielded Correlated Flowmeter, Tube Size 5
$607.00USD / Each
COMPARE
Water 20000 Shielded Correlated Flowmeter, Tube Size 6
$985.00USD / Each
COMPARE
Water 0.12 Unshielded Correlated Flowmeter Size Micro Tube
$267.00USD / Each
COMPARE
Water 2.3 Shielded Correlated Flowmeter Size. Micro Tube
$380.00USD / Each
COMPARE
Water 2.3 Shielded Correlated Flowmeter with Valve, Size 0 Tube
$939.00USD / Each
COMPARE
Water 8.6 Shielded Correlated Flowmeter with Valve, Size 1 shielded
$901.00USD / Each
COMPARE
Water 77 Shielded Correlated Flowmeter with Valve, Size 2 Tube
$849.00USD / Each
COMPARE
Water 640 Shielded Correlated Flowmeter with Valve, Size 3 Tube
$913.00USD / Each
COMPARE
Water 0.12 Shielded Correlated Flowmeter with Valve, Size 5 Tube
$1,496.00USD / Each
COMPARE
Water Shielded Correlated Flowmeter with Valve, Size Micro Tube
$829.00USD / Each
Showing 1 - 20 of 21