Cole-Parmer® Graduated Glass Cylinders, Class A, Calibrated to Deliver
Feel more confident about your aliquots - meet ISO 9000 requirements
  • Single metric scale
  • Class A cylinder glass meets ASTM E 438
  • Autoclavable
Choose cylinders with a Certificate of Graduation Accuracy or a NIST Calibration Report. Models with both Certificate of Graduation Accuracy and NIST Calibration Report feature fused-in serial numbers.
All support ISO 9000 requirements and are calibrated in accordance with ASTM E 542. Cylinders meet ASTM E 694 specifications and accuracy requirements of ASTM 1272. Each cylinder features an easy-to-read white scale and a molded base to decrease risk of accidentally knocking it over.
$12.00 - $150.00USD / Each
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Description
Availability Pricing
COMPARE
25 0.2 Graduated Cylinder with Certificate of Graduation Accuracy; 25 mL
$12.00USD / Each
COMPARE
25 0.2 Graduated Cylinder w/ Certificate of Graduation Accuracy, NIST Calibration; 25 mL
$16.00USD / Each
COMPARE
50 1 Graduated Cylinder with Certificate of Graduation Accuracy; 50 mL
$13.00USD / Each
COMPARE
50 1 Graduated Cylinder w/ Certificate of Graduation Accuracy, NIST Calibration; 50 mL
$27.00USD / Each
COMPARE
100 1 Graduated Cylinder with Certificate of Graduation Accuracy; 100 mL
$17.00USD / Each
COMPARE
100 1 Graduated Cylinder w/ Certificate of Graduation Accuracy, NIST Calibration; 100 mL
$31.00USD / Each
COMPARE
250 2 Graduated Cylinder with Certificate of Graduation Accuracy; 250 mL
$29.00USD / Each
COMPARE
250 2 Graduated Cylinder w/ Certificate of Graduation Accuracy, NIST Calibration; 250 mL
$56.00USD / Each
COMPARE
500 5 Graduated Cylinder with Certificate of Graduation Accuracy; 500 mL
$43.00USD / Each
COMPARE
500 5 Graduated Cylinder w/ Certificate of Graduation Accuracy, NIST Calibration; 500 mL
$76.00USD / Each
COMPARE
2000 20 Graduated Cylinder with Certificate of Graduation Accuracy; 2000 mL
$98.00USD / Each
COMPARE
2000 20 Graduated Cylinder w/ Certificate of Graduation Accuracy, NIST Calibration; 2000 mL
$150.00USD / Each
COMPARE
1000 10 Graduated Cylinder with Certificate of Graduation Accuracy; 1000 mL
$66.00USD / Each
COMPARE
1000 10 Graduated Cylinder w/ Certificate of Graduation Accuracy, NIST Calibration; 1000 mL
$114.00USD / Each
Showing 1- 14 of 14