Cole-Parmer® OVG-400 Series Gravity Convection Drying Ovens
Low energy consumption provides operating cost savings
  • Stainless steel inner chamber resist corrosion
  • Rounded inner chamber corners offer easy cleaning
  • Able to heat samples up to 392°F (200°C)
Gravity convection ovens feature a PID controller which provides you accurate and reliable temperature control up to 392°F (200°C). Natural convection heating allows sample temperature to be uniform; suitable for sterilization, powder drying, and high-temperature storage. The ovens require minimal heating time, reaching 212°F (100°C) in 15 to 30 minutes. Easy-to-view LCD reads time, current temperature, set temperature, running status, and fan status.
Silicone rubber gasket offers a tight seal, ensuring you good heat retention and low energy consumption. The gasket comes with a long service life and can be easily replaced when needed. Additional oven features include auto-start after power loss/return, temperature deviation alarm, overtemperature set point alarm, and overcurrent protection.
$869.50 - $1,946.00USD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (Liters) Max Temperature (° F) Power (VAC)
Availability Pricing
COMPARE
27 392 120
$869.50USD / Each
COMPARE
27 392 220
$869.50USD / Each
COMPARE
56 392 120
$1,041.00USD / Each
COMPARE
56 392 220
$1,041.00USD / Each
COMPARE
96 392 120
$1,382.00USD / Each
COMPARE
96 392 220
$1,382.00USD / Each
COMPARE
140 392 120
$1,662.00USD / Each
COMPARE
140 392 220
$1,662.00USD / Each
COMPARE
200 392 220
$1,946.00USD / Each
Showing 1- 9 of 9